Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL10RB Polyclonal Antibody
IL10RB Polyclonal Antibody
Supplier: Thermo Scientific PA580714
Description
IL10RB Polyclonal Antibody for Western Blot, ELISA

Specifications
IL10RB | |
Polyclonal | |
Unconjugated | |
IL10RB | |
6620401D04Rik; AI528744; CDW210B; CRF2-4; CRFB4; cytokine receptor class-II CRF2-4; cytokine receptor class-II member 4; Cytokine receptor family 2 member 4; cytokine receptor family II, member 4; D16H21S58; D21S58; D21S58h; D21S66; IL-10 receptor subunit beta; IL-10R subunit 2; IL-10R subunit beta; Il10r2; IL-10R2; IL10RB; IL-10RB; inte; interleukin 10 receptor 2; interleukin 10 receptor subunit beta; interleukin 10 receptor, beta; interleukin-10 receptor subunit 2; Interleukin-10 receptor subunit beta; RGD1560373 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3588 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with no preservative | |
Q08334 | |
IL10RB | |
Recombinant Human IL10RB/IL-10RB Protein (Met1-Ser220). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction