Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL11RA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119226
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119226 200 μL
PIPA5121831 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119226 Supplier Invitrogen™ Supplier No. PA5119226
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IL11RA Polyclonal Antibody for Western Blot, IHC (P), ICC/IF

Interleukin 11 is a stromal cell-derived cytokine that belongs to a family of pleiotropic and redundant cytokines that use the gp130 transducing subunit in their high affinity receptors. This gene encodes the IL-11 receptor, which is a member of the hematopoietic cytokine receptor family. This particular receptor is very similar to ciliary neurotrophic factor, since both contain an extracellular region with a 2-domain structure composed of an immunoglobulin-like domain and a cytokine receptor-like domain. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Jun 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL11RA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene IL11RA
Gene Accession No. Q14626, Q64385, Q99MF4
Gene Alias AI314697; CRSDA; Enhancer trap locus homolog 2; Etl2; etl-2; GP130; IL-11 receptor subunit alpha; IL-11 receptor subunit alpha-1; IL-11R subunit alpha; IL-11R subunit alpha-1; Il11ra; IL-11RA; Il11ra1; IL-11RA1; Il11ra2; Il-11ra-alpha; IL-11R-alpha; IL-11R-alpha-1; interleukin 11 receptor subunit alpha; interleukin 11 receptor, alpha; interleukin 11 receptor, alpha chain 1; interleukin 11 receptor, alpha chain 2; interleukin-11 receptor alpha chain; interleukin-11 receptor subunit alpha; Interleukin-11 receptor subunit alpha-1; locus 2; novel cytokine receptor 1; NR1; NR-1; sIL11RA
Gene Symbols IL11RA, Il11ra1
Host Species Rabbit
Immunogen Recombinant protein Interleukin 11 Receptor Alpha. The antigen corresponds to amino acid range 24-371 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16157, 245983, 3590
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.