Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL11RA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | IL11RA |
---|---|
Concentration | 0.55 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6161
![]() ![]() |
Proteintech Group Inc
102641AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Interleukin 11 is a stromal cell-derived cytokine that belongs to a family of pleiotropic and redundant cytokines that use the gp130 transducing subunit in their high affinity receptors. This gene encodes the IL-11 receptor, which is a member of the hematopoietic cytokine receptor family. This particular receptor is very similar to ciliary neurotrophic factor, since both contain an extracellular region with a 2-domain structure composed of an immunoglobulin-like domain and a cytokine receptor-like domain. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
IL11RA | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q14626, Q64385, Q99MF4 | |
16157, 245983, 3590 | |
IL11RA Fusion Protein Ag0373 | |
Primary | |
-20°C | |
IL11RA |
0.55 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
IL 11 receptor subunit alpha, IL 11R alpha, IL 11R subunit alpha, IL 11RA, IL11RA, interleukin 11 receptor, alpha | |
IL11RA, Il11ra1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title