Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL12B Antibody (I-1A4) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200054 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017628 0.05 mg
NB10064693 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017628 Supplier Novus Biologicals Supplier No. NB100646930.05MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL12B Monoclonal specifically detects IL12B in Human, Porcine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen IL12B
Applications Immunohistochemistry, Immunohistochemistry (Frozen), ELISA
Classification Monoclonal
Clone I-1A4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:1000
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Accession No. P29460
Gene Alias CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40
Gene Symbols IL12B
Host Species Mouse
Immunogen Recombinant IL-12
Purification Method Protein A or G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3593
Test Specificity Recognizes the p40 subunit of human interleukin-12, and binds to both the free subunit and the p35/p40 heterodimer. The reacts with both stimulated and unstimulated monocytes in immunohistology.
Target Species Pig, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.