Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL12B Antibody (I-1A4), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064693G
Description
IL12B Monoclonal specifically detects IL12B in Human, Porcine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
IL12B | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IL12B | |
Recombinant IL-12 | |
0.25 mL | |
Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
3593 | |
Human, Pig | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
I-1A4 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes the p40 subunit of human interleukin-12, and binds to both the free subunit and the p35/p40 heterodimer. The reacts with both stimulated and unstimulated monocytes in immunohistology. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction