Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL12B Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody

Supplier:  Novus Biologicals NB10093409

Catalog No. NB10093409


Only null left
Add to cart

Description

Description

IL12B Polyclonal specifically detects IL12B in Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

IL12B
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40
Goat
37 kDa
0.1 mg
Cell Cycle and Replication, Immunology, Innate Immunity
3593
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, ELISA
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000
P43432
IL12B
Peptide with sequence C-EARQQNKYENYS corresponding to internal region according to NP_032378.1.
Affinity Purified
RUO
Primary
Mouse, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only