Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL12B Antibody (OTI1A3), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271032AF647
Description
IL12B Monoclonal specifically detects IL12B in Human samples. It is validated for Western Blot.Specifications
IL12B | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IL12B | |
Full length recombinant protein of human IL12B (NP_002178) produced in SF9 cell. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI1A3 | |
Western Blot | |
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
3593.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction