Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL12B Antibody (OTI1A3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245391
Description
Ensure accurate, reproducible results in Western Blot
IL12B Monoclonal specifically detects IL12B in Human samples. It is validated for Western Blot.Specifications
IL12B | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 | |
Mouse | |
34.6 kDa | |
0.1 mL | |
Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
3593 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
OTI1A3 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
IL12B | |
Full length human recombinant protein of human IL12B (NP_002178) produced in SF9 cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction