Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL12RB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595976 Supplier Invitrogen™ Supplier No. PA595976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RTSECCFQDP PYPDADSGSA SGPRDLRCYR ISSDRYECSW QYEGPTAGVS HFLRCCLSSG RCCYFAAGSA TRLQFSDQAG VSVLYTVTLW VESWARNQTE KSPEVTLQLY NSVKYEPPLG DIKVSKLAGQ LRMEWETPDN QVGAEVQFRH RTPSSPWKLG DCGPQDDDTE SCLCPLEMNV AQEFQLRRRQ LGSQGSSWSK WSSPVCVPPE NPPQPQVRFS VEQLGQDGRR RLTLKEQPTQ LELPEGCQGL APGTEVTYRL QLHMLSCPCK AKATRTLHLG K; Positive Samples: A-431, MCF7, Mouse liver, Mouse thymus; Cellular Location: Membrane, Single-pass type I membrane protein.

The protein encoded by this gene is a type I transmembrane protein that belongs to the hemopoietin receptor superfamily. This protein binds to interleukin 12 with a low affinity, and is thought to be a part of IL12 receptor complex. This protein forms a disulfide-linked oligomer, which is required for its IL12 binding activity. The coexpression of this and IL12RB2 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The lack of expression of this gene was found to result in the immunodeficiency of patients with severe mycobacterial and Salmonella infections. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL12RB1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene IL12RB1
Gene Accession No. P42701, Q60837
Gene Alias CD212; cluster of differentiation 212; IL-12 receptor beta component; IL-12 receptor subunit beta-1; IL12R; IL-12R subunit beta-1; IL-12R[b]; IL12RB; Il12rb1; IL-12RB1; IL-12R-BETA1; IL-12R-beta-1; IMD30; interleukin 12 receptor subunit beta 1; interleukin 12 receptor, beta 1; interleukin-12 receptor beta-1 chain; Interleukin-12 receptor beta-1 chain precursor (IL-12R-beta1) (Interleukin-12 receptor beta) (IL-12 receptor beta component); interleukin-12 receptor subunit beta-1; MGC34454
Gene Symbols IL12RB1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 25-305 of human IL12RB1 (NP_7149121).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16161, 3594
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.