Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL12RB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521479
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Dog (80%), Pig (80%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a type I transmembrane protein identified as a subunit of the interleukin 12 receptor complex. The coexpression of this and IL12RB1 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The expression of this gene is up-regulated by interferon gamma in Th1 cells, and plays a role in Th1 cell differentiation. The up-regulation of this gene is found to be associated with a number of infectious diseases, such as Crohn's disease and leprosy, which is thought to contribute to the inflammatory response and host defense.
Specifications
IL12RB2 | |
Polyclonal | |
Unconjugated | |
IL12RB2 | |
A930027I18Rik; Ifnm; IL-12 receptor subunit beta-2; IL-12R subunit beta-2; Il12rb2; IL-12RB2; IL-12R-beta-2; interferon response to microorganisms; interleukin 12 receptor subunit beta 2; interleukin 12 receptor, beta 2; interleukin-12 receptor beta-2 chain; interleukin-12 receptor subunit beta-2; RP11-102M16.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3595 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q99665 | |
IL12RB2 | |
Recombinant protein encompassing a sequence within the center region of human IL12 Receptor beta2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction