Learn More
IL13 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | IL13 |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0870
![]() ![]() |
Rockland Immunochemicals
200401B55 |
100 μg |
Each for $636.24
|
|
|||||
50-199-0871
![]() ![]() |
Rockland Immunochemicals
200401B55S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for swine IL-13 protein. A BLAST analysis was used to suggest reactivity with IL-13 from swine sources based on 100% homology with the immunizing sequence. The following homologies may indicate chances of cross-reactivity when amino acid similarity and not just exact sequence is accounted: 89-90% homology to IL-13 from sheep, dolphin, beluga, and bovine, 83-88% to human and certain monkey IL-13, 80-82% to dusky titi, macaque, baboon, mangabey, dog, llama, camel, and galago, 75-78% to horse, and 67-72% to rat, mouse, and gerbil. Cross-reactivity with IL-13 from other sources has not been determined.
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.Specifications
IL13 | |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Bovine, Ovine, Pig | |
Q95J68, Q9XSV9 | |
281247, 396721, 664707 | |
This protein-A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant protein raised in yeast, corresponding to mature swine IL-13 protein. | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Il13 |
1 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
ALRH; ALRH antibody; BHR1; BOS_7773; Il13; Il-13; ILN; Interleukin; interleukin 13; Interleukin13; Interleukin-13; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
Il13 | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.