Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL17A Mouse anti-Human, Clone: 3-36.2.7.3, Abnova™

Catalog No. 89035504 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-504 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-504 Supplier Abnova Corporation Supplier No. MAB14106

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against recombinant human IL17A.

  • High specificity
  • High reactivity

Specifications

Antigen IL17A
Applications ELISA
Classification Monoclonal
Clone 3-36.2.7.3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant human IL17A.
Dilution ELISA (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS.
Gene Accession No. Q16552
Gene Alias CTLA8/IL-17/IL-17A/IL17
Gene Symbols IL17A
Host Species Mouse
Immunogen Recombinant protein corresponding to human IL17A.
Purification Method Protein G affinity purification
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3605
Test Specificity This antibody has successfully been used as the coating antibody in a sandwich ELISA.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.