Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL17A Mouse anti-Human, Clone: 4K5F6, Abnova™

Catalog No. 89934362 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-362 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-362 Supplier Abnova Corporation Supplier No. MAB12727
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human IL17A.

The protein encoded by this gene is a proinflammatory cytokine produced by activated T cells. This cytokine regulates the activities of NF-kappaB and mitogen-activated protein kinases. This cytokine can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of this cytokine are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis. [provided by RefSeq]

Specifications

Antigen IL17A
Applications Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 4K5F6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human IL17A.
Dilution Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) Western Blot (0.2-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide).
Gene Alias CTLA8/IL-17/IL-17A/IL17
Gene Symbols IL17A
Host Species Mouse
Immunogen A peptide corresponding to amino acids 1-75 of human IL17A.
Purification Method Protein G purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3605
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.