Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL17B Monoclonal Antibody (OTI8F7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811134

Description
The protein encoded by this gene is a T cell-derived cytokine that shares sequence similarity with IL17. This cytokine was reported to stimulate the release of TNF alpha (TNF) and IL1 beta (IL1B) from a monocytic cell line. Immunohistochemical analysis of several nerve tissues indicated that this cytokine is primarily localized to neuronal cell bodies.Specifications
IL17B | |
Monoclonal | |
Unconjugated | |
IL17B | |
IL-17B, IL-20, NIRF, ZCYTO7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
27190 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
OTI8F7 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UHF5 | |
IL17B | |
Full length human recombit protein of human IL17B produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction