Learn More
IL17B Mouse anti-Human, Clone: OTI1F10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811129

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a T cell-derived cytokine that shares sequence similarity with IL17. This cytokine was reported to stimulate the release of TNF alpha (TNF) and IL1 beta (IL1B) from a monocytic cell line. Immunohistochemical analysis of several nerve tissues indicated that this cytokine is primarily localized to neuronal cell bodies.Specifications
IL17B | |
Monoclonal | |
Unconjugated | |
IL17B | |
IL-17B, IL-20, NIRF, ZCYTO7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
27190 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1F10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UHF5 | |
IL17B | |
Full length human recombit protein of human IL17B produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.