Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL17C Rabbit anti-Human, Non-human Primate, Polyclonal, FabGennix

Catalog No. 502207426
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7426 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7426 Supplier Fabgennix Inc Supplier No. IL17RC301AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL17C Polyclonal antibody specifically detects IL17C in Human, Non-human Primate samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen IL17C
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene IL17C
Gene Accession No. Q9P0M4
Gene Alias CX2; cytokine CX2; Il17c; IL-17C; IL-21; interleukin 17C; interleukin-17C; UNQ561/PRO1122
Host Species Rabbit
Immunogen Synthetic peptide corresponding to IL17RC protein common to C1, C2, C5, C8, C9, C11, C12 isoforms.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27189, 468067
Target Species Human, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.