Learn More
Invitrogen™ IL17RB Polyclonal Antibody

Description
Immunogen sequence: REPTVQCGSE TGPSPEWMLQ HDLIPGDLRD LRVEPVTTSV ATGDYSILMN VSWVLRADAS IRLLKATKIC VTGKSNFQSY SCVRCNYTEA FQTQTRPSGG KWTFSYIGFP VELNTVYFIG AHNIPNANMN EDGPSMSVNF TSPGCLDHIM KYKKKCVKAG SLWDPNITAC KKNEETVEVN FTTTPLGNRY MALIQHSTII GFSQVFEPHQ KKQTRASVVI PVTGDSEGAT VQLTPYFPTC GSDCIRHKGT VVLCPQTGVP FPLDNNKSKP GGWLP; Positive Samples: HepG2, 293T, SW620, A-549, B-cell, Mouse liver, Rat liver; Cellular Location: Cell membrane, Single-pass type I membrane protein.
Specifications
Specifications
| Antigen | IL17RB |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | IL17RB |
| Gene Accession No. | Q9JIP3, Q9NRM6 |
| Gene Alias | CRL4; cytokine receptor CRL4; cytokine receptor-like 4; ecotropic viral integration site 27; EVI27; IL-17 receptor B; IL-17 receptor homolog 1; IL-17B; IL-17B receptor; IL17BR; IL-17ER; Il17rb; IL-17RB; IL17RH1; IL-17Rh1; IL-20; interleukin 17 receptor B; interleukin 17 receptor homolog 1; interleukin 17B receptor; interleukin-17 receptor B; Interleukin-17B receptor; MGC138900; MGC138901; MGC5245; UNQ2501/PRO19612; ZCYTO7 |
| Gene Symbols | IL17RB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.