Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL17RC Isoform 1 Precursor Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102004
Description
Antibody detects endogenous levels of total IL17RC.
This gene encodes a single-pass type I membrane protein that shares similarity with the interleukin-17 receptor (IL-17RA). Unlike IL-17RA, which is predominantly expressed in hemopoietic cells, and binds with high affinity to only IL-17A, this protein is expressed in nonhemopoietic tissues, and binds both IL-17A and IL-17F with similar affinities. The proinflammatory cytokines, IL-17A and IL-17F, have been implicated in the progression of inflammatory and autoimmune diseases. Multiple alternatively spliced transcript variants encoding different isoforms have been detected for this gene, and it has been proposed that soluble, secreted proteins lacking transmembrane and intracellular domains may function as extracellular antagonists to cytokine signaling.
Specifications
IL17RC Isoform 1 Precursor | |
Polyclonal | |
Unconjugated | |
IL17RC | |
CANDF9; IL-17 receptor C; IL17F receptor; IL17RC; IL17Rhom; IL17RL; IL-17RL; IL17-RL; interleukin 17 receptor C; interleukin-17 receptor C; Interleukin-17 receptor homolog; interleukin-17 receptor-like protein; UNQ6118/PRO20040/PRO38901; ZcytoR14 | |
Rabbit | |
Affinity chromatography | |
RUO | |
84818 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8NAC3 | |
IL17RC | |
A synthesized peptide derived from human IL17RC(Accession Q8NAC3), corresponding to amino acid residues L717-F767. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction