Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL17RC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119227
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119227 200 μL
PIPA5121828 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119227 Supplier Invitrogen™ Supplier No. PA5119227
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IL17RC Polyclonal Antibody for Western Blot

This gene encodes a single-pass type I membrane protein that shares similarity with the interleukin-17 receptor (IL-17RA). Unlike IL-17RA, which is predominantly expressed in hemopoietic cells, and binds with high affinity to only IL-17A, this protein is expressed in nonhemopoietic tissues, and binds both IL-17A and IL-17F with similar affinities. The proinflammatory cytokines, IL-17A and IL-17F, have been implicated in the progression of inflammatory and autoimmune diseases. Multiple alternatively spliced transcript variants encoding different isoforms have been detected for this gene, and it has been proposed that soluble, secreted proteins lacking transmembrane and intracellular domains may function as extracellular antagonists to cytokine signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL17RC
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene IL17RC
Gene Accession No. Q8NAC3
Gene Alias 1110025H02Rik; CANDF9; Gm19850; IL-17 receptor C; IL17F receptor; IL17RC; IL-17RC; IL17-RC; IL17Rhom; Il17rl; IL-17RL; IL17-RL; interleukin 17 receptor C; interleukin-17 receptor C; Interleukin-17 receptor homolog; interleukin-17 receptor-like protein; UNQ6118/PRO20040/PRO38901; ZcytoR14
Gene Symbols IL17RC
Host Species Rabbit
Immunogen Recombinant protein Interleukin 17 Receptor C. The antigen corresponds to amino acid range 257-477 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100152641, 84818
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.