Learn More
Invitrogen™ IL17RD Polyclonal Antibody

Description
Immunogen sequence: LACKPFWKPR NLNISQHGSD MQVSFDHAPH NFGFRFFYLH YKLKHEGPFK RKTCKQEQTT ETTSCLLQNV SPGDYIIELV DDTNTTRKVM Highest antigen sequence indentity to the following orthologs: Rat - 82%, Mouse - 84%.
Specifications
Specifications
| Antigen | IL17RD |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Il17rd |
| Gene Accession No. | Q8NFM7 |
| Gene Alias | 2810004A10Rik; AI428510; HH18; hSef; IL-17 receptor D; IL17RD; IL-17RD; IL17Rhom; IL17RLM; interleukin 17 receptor D; interleukin-17 receptor D; Interleukin-17 receptor-like protein; mSef; RGD1565237; SEF; sef homolog; Sef-S; UNQ6115/PRO20026 |
| Gene Symbols | Il17rd |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.