Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL18 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501726491 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6491 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6491 Supplier Proteintech Group Inc Supplier No. 106631AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody 10663-1-AP is raised against the full length IL18 precursor, and it recognizes both the pre-IL18 and mature IL18 theoretically.

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.

Specifications

Antigen IL18
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene IL18
Gene Accession No. Q14116
Gene Alias Iboctadekin, IFN gamma inducing factor, IGIF, IL 1 gamma, IL 18, IL 1g, IL18, IL1F4, Interleukin 1 gamma, Interleukin 18, Mature IL18, pre IL18, Pre-IL18
Gene Symbols IL18
Host Species Rabbit
Immunogen IL18 Fusion Protein Ag1063
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3606
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.