Learn More
Invitrogen™ IL18BP Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
Supplier: Invitrogen™ 500P153GBT25UG

Description
Preparation: Produced from sera of goats immunized with highly pure Recombinant Murine IL-18BP. Anti-Murine IL-18BP-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Murine IL-18BP by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine IL-18BP (500-P153G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine IL-18BP. Western Blot: To detect mIL-18BP by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mIL-18BP is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P153GBT-1MG will be provided as 2 x 500 μg
Interleukin-18 binding protein (IL-18BP) is a constitutively secreted glycoprotein that acts as a natural antagonist for interleukin-18 (IL-18) by preventing interaction between IL-18 and its receptors. Through this binding, IL-18BP neutralizes the proinflammatory role of IL-18 in cell-mediated immune responses, resulting in reduced IFN-gamma production by T helper type I cells and inhibition of allospecific cytotoxic T lymphocyte (CTL) activity through augmented natural killer (NK) cell cytotoxicity. As an inhibitor of IL-18 and an essential component of skin homeostasis, IL-18BP has been linked to a variety of diseases, including eczema, asthma, and the autoimmune condition, Crohn's disease. The upregulation of IL-18BP by keratinocytes expressing the human papillomavirus (HPV) E7 oncoprotein has been shown to decrease IL-18-induced IFN-gamma production and IL-18-mediated T cell activation. Elevated levels of IL-18 and IL-18BP, as well as proinflammatory cytokines IFN-gamma, TNF-alpha, IL-1beta and IL-8, have been linked to chronic inflammation and observed in macrophages (including Kupffer cells) isolated from lesions and intestinal tissues of Crohn's disease patients. As a member of the immunoglobulin-like class of receptors, IL-18BP contains a single immunoglobulin (Ig) domain; although two (b and d) of the four identified human isoforms (a-d) contain incomplete domains that greatly reduce their binding affinity.
Specifications
IL18BP | |
Polyclonal | |
Biotin | |
IL18BP | |
Igifbp; Il18bp; IL-18BP; IL18BPa; interferon gamma inducing factor binding protein; interferon gamma-inducing factor-binding protein; interleukin 18 binding protein; interleukin-18 binding protein; interleukin-18-binding protein; MC51L-53L-54L homolog gene product; MC54L; Tadekinig-alfa | |
Goat | |
Antigen Affinity Chromatography | |
RUO | |
16068 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
0.1-1.0 mg/mL | |
PBS with no preservative | |
Q9Z0M9 | |
IL18BP | |
E.coli-derived Recombinant Murine IL-18BP. | |
25 μg | |
Primary | |
Mouse | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.