Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL18BPd Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547097
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547097 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547097 Supplier Invitrogen™ Supplier No. PA547097
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, approximately 25% cross-reactivity with recombinant mouse IL-18 BPc is observed and less than 2% cross-reactivity with recombinant human IL-18 BPa is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL18BPd
Applications ELISA, Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Il18bp
Gene Accession No. Q9Z0M9
Gene Alias Igifbp; Il18bp; IL-18BP; interferon gamma inducing factor binding protein; interferon gamma-inducing factor-binding protein; interleukin 18 binding protein; interleukin-18-binding protein; MC54L
Gene Symbols Il18bp
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-18 BPd Thr27-Ala191.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16068
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.