Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL18R1 Antibody (006), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290417PE
Description
IL18R1 Monoclonal antibody specifically detects IL18R1 in Mouse samples. It is validated for ELISA, ELISASpecifications
IL18R1 | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
006 | |
ELISA, Sandwich ELISA Capture | |
CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 | |
Rabbit immunized with purified, recombinant Mouse IL18R1 (Accession#: NP_032391.1, Met1-Gly326) | |
0.1 mL | |
CD Markers | |
8809 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction