Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL18R1 Antibody (11), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289797H
Description
IL18R1 Monoclonal specifically detects IL18R1 in Human samples. It is validated for ELISA.Specifications
IL18R1 | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
8809 | |
Human | |
Purified |
ELISA | |
11 | |
ELISA | |
CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL18R1 (NP_003846.1; Met 1-Arg 329). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Mouse IL18R1/ IL1RRP/ CD218a Mouse SIGIRR Human IL1R1/ CD121a Human IL1R2/ IL1RB/ CD121b Human IL1R3/ IL1RAP Human IL1R8/ IL1RAPL1 Human IL1RL1/ IL1R4/ ST2 Human IL18RAP/ IL1R7 Human IL1R9/ IL1RAPL2 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction