Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL18R1 Antibody (11), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289797JF646
Description
IL18R1 Monoclonal antibody specifically detects IL18R1 in Human samples. It is validated for ELISA, ELISASpecifications
IL18R1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Sandwich ELISA | |
11 | |
ELISA, Sandwich ELISA Capture | |
CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL18R1 (Accession#: NP_003846.1; Met 1-Arg 329). | |
0.1 mL | |
CD Markers | |
8809 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction