Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL18R1 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298223F
Description
IL18R1 Polyclonal specifically detects IL18R1 in Mouse samples. It is validated for ELISA.Specifications
IL18R1 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
8809 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 | |
Produced in rabbits immunized with purified, recombinant IL18R1 (NP_032391.1; Met 1-Gly 326). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction