Learn More
Invitrogen™ IL18R1 Monoclonal Antibody (112624)

Description
In direct ELISAs, this antibody does not cross-react with recombinant mouse (rm) IL-1 RI, rmIL-1 RII, rhIL-1 R3, recombinant human (rh) IL-1 R4, rhIL-1 R5, recombinant rat IL-1 R6, rmIL-1 R7, rhIL-1 R8, rhIL-1 R9, or rmSIGIRR. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
Specifications
| Antigen | IL18R1 |
| Applications | Neutralization, Western Blot |
| Classification | Monoclonal |
| Clone | 112624 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Il18r1 |
| Gene Accession No. | Q61098 |
| Gene Alias | CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.