Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL18R1 Recombinant Rabbit Monoclonal Antibody (3Q2D5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551403
Description
Immunogen sequence: ENSYYQTLVN STSLYKNCKK LLLENNKNPT IKKNAEFEDQ GYYSCVHFLH HNGKLFNITK TFNITIVEDR SNIVPVLLGP KLNHVAVELG KNVRLNCSAL LNEEDVIYWM FGEENGSDPN IHEEKEMRIM TPEGKWHASK VLRIENIGES NLNVLYNCTV ASTGGTDTKS FILVRKADMA.
IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
Specifications
IL18R1 | |
Recombinant Monoclonal | |
1.1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q13478, Q61098 | |
Il18r1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 141-320 of human IL18R1/IL-18 R alpha (NP_003846.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Liquid |
ELISA, Western Blot | |
3Q2D5 | |
Unconjugated | |
Il18r1 | |
CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha | |
Rabbit | |
Affinity chromatography | |
RUO | |
16182, 8809 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction