Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL18RAP Recombinant Rabbit Monoclonal Antibody (122)
GREENER_CHOICE

Catalog No. PIMA530551
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530551 Supplier Invitrogen™ Supplier No. MA530551
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human IL18RAP.

The protein encoded by this gene is an accessory subunit of the heterodimeric receptor for IL18. This protein enhances the IL18 binding activity of IL18R1 (IL1RRP), a ligand binding subunit of IL18 receptor. The coexpression of IL18R1 and this protein is required for the activation of NF-kappaB and MAPK8 (JNK) in response to IL18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL18RAP
Applications ELISA
Classification Recombinant Monoclonal
Clone 122
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IL18RAP
Gene Accession No. O95256
Gene Alias Accessory protein-like; ACPL; AcPL accessory protein-like); CD218 antigen-like family member B; CD218b; CDw218b; cluster of differentiation w218b; IL-18 RA; IL-18 receptor accessory protein; IL-18 receptor beta; IL-18RAcP; IL18RAP; IL18RB; IL-18Rbeta; IL-18R-beta; IL-1R accessory protein-like; IL1R7; IL-1R7; IL-1R-7; IL-1RAcPL; interleukin 18 receptor accessory protein; interleukin-1 receptor 7; Interleukin-18 receptor accessory protein; Interleukin-18 receptor accessory protein-like; Interleukin-18 receptor beta
Gene Symbols IL18RAP
Host Species Rabbit
Immunogen Recombinant Human IL18RAP Protein (Met1-Gly357).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8807
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.