Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL1A Mouse anti-Human, Clone: , Abnova™

Catalog No. 89934365 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-365 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-365 Supplier Abnova Corporation Supplier No. MAB12516
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human IL1A.

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. This cytokine is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury, and thus induces apoptosis. This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. It has been suggested that the polymorphism of these genes is associated with rheumatoid arthritis and Alzheimer's disease. [provided by RefSeq]

Specifications

Antigen IL1A
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human IL1A.
Dilution ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:400) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide).
Gene Alias IL-1A/IL1/IL1-ALPHA/IL1F1
Gene Symbols IL1A
Host Species Mouse
Immunogen Recombinant protein corresponding to human IL1A.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3552
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.