Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Il1a, Mouse anti-Mouse, Clone: Bamboo-2, Abnova™

Catalog No. 89998860 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-860 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-860 Supplier Abnova Corporation Supplier No. MAB16116
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen Il1a
Applications ELISA
Classification Monoclonal
Clone Bamboo-2
Conjugate Unconjugated
Dilution ELISA (as a detection antibody) The optimal working dilution should be determined by the end user.
Formulation In PBS (10% glycerol, 0.02% sodium azide).
Gene interleukin 1 alpha
Gene Alias Il-1a
Gene Symbols Il1a
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse mature Il1a.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16175
Target Species Mouse
Content And Storage For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.