Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL1F10 (IL-38) Mouse anti-Human, Clone: H127C, MilliporeSigma™

Catalog No. MABF2754B10 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2754B10 100 μg
MABF2754B25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2754B10 Supplier MilliporeSigma Supplier No. MABF2754B100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL1F10 (IL-38) Monoclonal antibody specifically detects IL1F10 (IL-38) in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1F10 (IL-38)
Applications ELISA
Classification Monoclonal
Clone H127C
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in PBS, 1.5% BSA with 0.1% sodium azide.
Host Species Mouse
Immunogen His-tagged full-length recombinant human Interleukin-1 family member 10 (IL1F10).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.