Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL1F10 Mouse anti-Human, Clone: 5B2F5, Proteintech
SDP

Catalog No. 501736409 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6409 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6409 Supplier Proteintech Group Inc Supplier No. 603191IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The cytokine IL-1 is responsible for initiating a variety of activities through the activation of transcription factors, NF-kappa B and AP-1, thereby promoting host response to injury or infection. IL-1F10 (Interleukin-1 family member 10), also known as FIL1T, IL1HY2 or FKSG75, is highly expressed in the skin, spleen and proliferating B-cells of the tonsil. IL-1F10 binds soluble IL-1 receptor type 1, and is one of nine Interleukin 1 families clustered on chromosome 2, where it is thought to participate in the regulation of adapted and innate immune responses. IL-1F10 and IL-1RN polymorphisms may play an important role in the susceptibility to developing rheumatoid arthritis.

Specifications

Antigen IL1F10
Applications Western Blot
Classification Monoclonal
Clone 5B2F5
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IL1F10
Gene Accession No. Q8WWZ1
Gene Alias FIL1 theta, FIL1T, FKSG75, IL 1 theta, IL 1F10, IL 1HY2, IL1 theta, IL1F10, IL1HY2, Interleukin 1 family member 10, Interleukin 1 HY2, Interleukin 1 theta, UNQ6119/PRO20041
Gene Symbols IL1F10
Host Species Mouse
Immunogen IL1F10 Fusion Protein Ag17250
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84639
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.