Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL1F5 Monoclonal Antibody (04), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA542975
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542975 100 μL
PIMA543519 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA542975 Supplier Invitrogen™ Supplier No. MA542975
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human IL-1F5

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine was shown to specifically inhibit the activation of NF-kappaB induced by interleukin 1 family, member 6 (IL1F6). This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Two alternatively spliced transcript variants encoding the same protein have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1F5
Applications ELISA
Classification Monoclonal
Clone 04
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IL36RN
Gene Accession No. Q9UBH0
Gene Alias AI413231; FIL1; FIL1 delta; FIL1(DELTA); FIL1D; Fil1delta; IL-1 delta; IL-1 related protein 3; Il1f5; IL-1F5; IL1F5 (Canonical product IL-1F5a); IL-1F5 (IL-1HY1, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta); Il1h3; Il-1h3; IL1HY1; IL-1HY1; IL1L1; IL-1L1; IL-1ra homolog 1; IL-1-related protein 3; IL1RP3; IL-1RP3; IL36RA; IL-36Ra; IL36RN; Interleukin; interleukin 1 family, member 5; interleukin 1 family, member 5 (delta); interleukin 1 receptor antagonist homolog 1; interleukin 36 receptor antagonist; interleukin-1 delta; Interleukin-1 family member 5; interleukin-1 family member 5; interleukin-36 receptor antagonist protein; interleukin-1 homolog 3; Interleukin-1 HY1; interleukin-1 receptor antagonist homolog 1; Interleukin-1-like protein 1; interleukin-36 receptor antagonist protein; MGC29840; PSORP; PSORS14; UNQ1896/PRO4342
Gene Symbols IL36RN
Host Species Mouse
Immunogen Recombinant Human IL-1F5 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26525
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.