Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL1R1 Monoclonal Antibody (D14e3)
GREENER_CHOICE

Catalog No. FERMA549052
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549052 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549052 Supplier Invitrogen™ Supplier No. MA549052
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL1R1 Monoclonal Antibody for Western Blot

The IL-1 system includes two agonists (IL-1 and IL-1), converting enzymes, antagonists, two receptors (IL-1 RI and IL-1 RII) and the IL-1 receptor accessory protein. Interleukin-1 signal is transduced through the type I receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1R1
Applications Western Blot
Classification Monoclonal
Clone D14e3
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene Il1r1
Gene Accession No. P13504
Gene Alias antigen CD121a; CD121 alpha; CD121 antigen-like family member A; CD121a; CD121b; D2S1473; il1 receptor; il-1 receptor; IL-1 receptor alpha chain; IL1R; il-1r; IL1R1; IL-1R1; IL-1R-1; Il1r-1; IL1RA; IL-1R-alpha; IL1RT1; IL-1RT1; IL-1RT-1; IL-iR; interleukin 1 beta receptor type 1 short transmembrane form; interleukin 1 beta receptor type 1 soluble form; interleukin 1 bete receptor type 1 long transmembrane form; interleukin 1 receptor alpha, type I; interleukin 1 receptor type 1; interleukin 1 receptor, type I; interleukin-1 receptor; interleukin-1 receptor alpha; Interleukin-1 receptor type 1; Interleukin-1 receptor type 1, membrane form; Interleukin-1 receptor type 1, soluble form; Interleukin-1 receptor type I; mIL-1R1; p80; sIL-1R1
Gene Symbols Il1r1
Host Species Rat
Immunogen The monoclonal antibody binds to soluble mouse interleukin-1 receptor type I.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16177
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.