Learn More
Invitrogen™ IL1R1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547937
Description
In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse (rm) IL-18 R is observed and 5% cross-reactivity with rmIL-18 R beta, recombinant human IL-1 RI, and recombinant rat IL-1 Rrp2 is observed and less than 1% cross-reactivity with rmIL-1 RII is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
The IL-1 system includes two agonists (IL-1 and IL-1), converting enzymes, antagonists, two receptors (IL-1 RI and IL-1 RII) and the IL-1 receptor accessory protein. Interleukin-1 signal is transduced through the type I receptor.
Specifications
| IL1R1 | |
| Polyclonal | |
| Unconjugated | |
| Il1r1 | |
| antigen CD121a; CD121 alpha; CD121 antigen-like family member A; CD121a; CD121b; D2S1473; il1 receptor; il-1 receptor; IL-1 receptor alpha chain; IL1R; il-1r; IL1R1; IL-1R1; IL-1R-1; Il1r-1; IL1RA; IL-1R-alpha; IL1RT1; IL-1RT1; IL-1RT-1; IL-iR; interleukin 1 beta receptor type 1 short transmembrane form; interleukin 1 beta receptor type 1 soluble form; interleukin 1 bete receptor type 1 long transmembrane form; interleukin 1 receptor alpha, type I; interleukin 1 receptor type 1; interleukin 1 receptor, type I; interleukin-1 receptor; interleukin-1 receptor alpha; Interleukin-1 receptor type 1; Interleukin-1 receptor type 1, membrane form; Interleukin-1 receptor type 1, soluble form; Interleukin-1 receptor type I; mIL-1R1; p80; sIL-1R1 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 16177 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P13504 | |
| Il1r1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-1 RI Leu20-Lys338. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.