Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL1R1 Recombinant Rabbit Monoclonal Antibody (191), APC
GREENER_CHOICE

Catalog No. PIMA540958
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA540958 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA540958 Supplier Invitrogen™ Supplier No. MA540958
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.

The IL-1 system includes two agonists (IL-1 and IL-1), converting enzymes, antagonists, two receptors (IL-1 RI and IL-1 RII) and the IL-1 receptor accessory protein. Interleukin-1 signal is transduced through the type I receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1R1
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 191
Concentration 0.1 mg/mL
Conjugate APC
Formulation PBS with 0.5% BSA and 0.03% ProClin 300
Gene Il1r1
Gene Accession No. P13504
Gene Alias antigen CD121a; CD121 alpha; CD121 antigen-like family member A; CD121a; CD121b; D2S1473; il1 receptor; il-1 receptor; IL-1 receptor alpha chain; IL1R; il-1r; IL1R1; IL-1R1; IL-1R-1; Il1r-1; IL1RA; IL-1R-alpha; IL1RT1; IL-1RT1; IL-1RT-1; IL-iR; interleukin 1 beta receptor type 1 short transmembrane form; interleukin 1 beta receptor type 1 soluble form; interleukin 1 bete receptor type 1 long transmembrane form; interleukin 1 receptor alpha, type I; interleukin 1 receptor type 1; interleukin 1 receptor, type I; interleukin-1 receptor; interleukin-1 receptor alpha; Interleukin-1 receptor type 1; Interleukin-1 receptor type 1, membrane form; Interleukin-1 receptor type 1, soluble form; Interleukin-1 receptor type I; mIL-1R1; p80; sIL-1R1
Gene Symbols Il1r1
Host Species Rabbit
Immunogen Recombinant Mouse IL-1R1/CD121a protein.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16177
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.