Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL1R1 Recombinant Rabbit Monoclonal Antibody (007)
GREENER_CHOICE

Catalog No. PIMA529867
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529867 Supplier Invitrogen™ Supplier No. MA529867
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Rat IL-1R1/CD121a.

The IL-1 system includes two agonists (IL-1 and IL-1), converting enzymes, antagonists, two receptors (IL-1 RI and IL-1 RII) and the IL-1 receptor accessory protein. Interleukin-1 signal is transduced through the type I receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1R1
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Clone 007
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Il1r1
Gene Accession No. Q02955
Gene Alias antigen CD121a; CD121 alpha; CD121 antigen-like family member A; CD121a; CD121b; D2S1473; il1 receptor; il-1 receptor; IL-1 receptor alpha chain; IL1R; il-1r; IL1R1; IL-1R1; IL-1R-1; Il1r-1; IL1RA; IL-1R-alpha; IL1RT1; IL-1RT1; IL-1RT-1; IL-iR; interleukin 1 beta receptor type 1 short transmembrane form; interleukin 1 beta receptor type 1 soluble form; interleukin 1 bete receptor type 1 long transmembrane form; interleukin 1 receptor alpha, type I; interleukin 1 receptor type 1; interleukin 1 receptor, type I; interleukin-1 receptor; interleukin-1 receptor alpha; Interleukin-1 receptor type 1; Interleukin-1 receptor type 1, membrane form; Interleukin-1 receptor type 1, soluble form; Interleukin-1 receptor type I; mIL-1R1; p80; sIL-1R1
Gene Symbols Il1r1
Host Species Rabbit
Immunogen Recombinant Rat IL-1R1/CD121a protein (Met1-Lys352).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25663
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.