Learn More
Invitrogen™ IL1R2 Monoclonal Antibody (34141), APC

Description
When used in combination with the biotinylated human IL-1 RII affinity purified polyclonal detection antibody in sandwich ELISAs, no significant cross-reactivity or interference was observed with recombinant human (rh) IL-1ra, rhIL-1 RI, recombinant mouse IL-1ra, or recombinant rat IL-1ra.
Specifications
Specifications
| Antigen | IL1R2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 34141 |
| Conjugate | APC |
| Formulation | Buffered saline solution with ≤ 0.5% BSA and 0.09% sodium azide |
| Gene | Il1r2 |
| Gene Accession No. | P27930 |
| Gene Alias | antigen CDw121b; CD121 antigen-like family member B; CD121b; CDw121b; IL-1 receptor beta chain; IL-1 type II receptor; Il1r2; Il-1r2; IL-1R-2; Il1r-2; IL1R2c; IL1RB; IL-1R-beta; IL-1RT2; IL-1RT-2; ILR2; interleukin 1 receptor type 2; interleukin 1 receptor type II variant 3; interleukin 1 receptor, type II; interleukin-1 receptor beta; Interleukin-1 receptor type 2; Interleukin-1 receptor type 2, membrane form; Interleukin-1 receptor type 2, soluble form; Interleukin-1 receptor type II; mIL-1R2; sIL-1R2; type II interleukin-1 receptor, beta |
| Gene Symbols | Il1r2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.