Learn More
Invitrogen™ IL1R2 Polyclonal Antibody

Description
Immunogen sequence: GALWLLPALQ EDSGTYVCTT RNASYCDKMS IELRVFENTD AFLPFISYPQ ILTLSTSGVL VCPDLSEFTR DKTDVKIQWY KDSLLLDK Highest antigen sequence indentity to the following orthologs: Mouse - 64%, Rat - 64%.
Specifications
Specifications
| Antigen | IL1R2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Il1r2 |
| Gene Accession No. | P27930 |
| Gene Alias | antigen CDw121b; CD121 antigen-like family member B; CD121b; CDw121b; IL-1 receptor beta chain; IL-1 type II receptor; Il1r2; Il-1r2; IL-1R-2; Il1r-2; IL1R2c; IL1RB; IL-1R-beta; IL-1RT2; IL-1RT-2; ILR2; interleukin 1 receptor type 2; interleukin 1 receptor type II variant 3; interleukin 1 receptor, type II; interleukin-1 receptor beta; Interleukin-1 receptor type 2; Interleukin-1 receptor type 2, membrane form; Interleukin-1 receptor type 2, soluble form; Interleukin-1 receptor type II; mIL-1R2; sIL-1R2; type II interleukin-1 receptor, beta |
| Gene Symbols | Il1r2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.