Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL1R2 Recombinant Rabbit Monoclonal Antibody (041)
GREENER_CHOICE

Catalog No. PIMA530559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530559 Supplier Invitrogen™ Supplier No. MA530559
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human IL-1R2/CD121b.

The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein binds interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA), and acts as a decoy receptor that inhibits the activity of its ligands. Interleukin 4 (IL4) is reported to antagonize the activity of interleukin 1 by inducing the expression and release of this cytokine. This gene and three other genes form a cytokine receptor gene cluster on chromosome 2q12. Two alternatively spliced transcript variants encoding the same protein have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1R2
Applications ELISA
Classification Recombinant Monoclonal
Clone 041
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Il1r2
Gene Accession No. P27930
Gene Alias antigen CDw121b; CD121 antigen-like family member B; CD121b; CDw121b; IL-1 receptor beta chain; IL-1 type II receptor; Il1r2; Il-1r2; IL-1R-2; Il1r-2; IL1R2c; IL1RB; IL-1R-beta; IL-1RT2; IL-1RT-2; ILR2; interleukin 1 receptor type 2; interleukin 1 receptor type II variant 3; interleukin 1 receptor, type II; interleukin-1 receptor beta; Interleukin-1 receptor type 2; Interleukin-1 receptor type 2, membrane form; Interleukin-1 receptor type 2, soluble form; Interleukin-1 receptor type II; mIL-1R2; sIL-1R2; type II interleukin-1 receptor, beta
Gene Symbols Il1r2
Host Species Rabbit
Immunogen Recombinant Human IL-1R2/CD121b Protein (Met1-Glu343).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7850
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.