Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL1RA Monoclonal Antibody (02)
GREENER_CHOICE

Catalog No. PIMA529352
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529352 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529352 Supplier Invitrogen™ Supplier No. MA529352
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IL-1RA.

The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1RA
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IL1RN
Gene Accession No. P25085, P25086
Gene Alias Anakinra; CD121A; cytokine; D2S1473; DIRA; F630041P17Rik; ICIL-1RA; IL 1 receptor antagonist; IL1 inhibitor; il1 receptor; il-1 receptor; IL1 receptor antagonist; IL1F3; IL1R; il-1r; IL1RA; IL-1RA; IL1-RA; IL-1ra3; IL-1R-alpha; IL1RN; IL-1RN; IL1RN (IL1F3); interleukin 1 receptor antagonist; interleukin-1 receptor; interleukin-1 receptor antagonist; interleukin-1 receptor antagonist protein; interleukin-1 receptor antagonist secretory form; intracellular IL-1 receptor antagonist type II; intracellular interleukin-1 receptor antagonist (icIL-1ra); IRAP; IRAP1; MGC10430; MVCD4; P80; type II interleukin-1 receptor antagonist
Gene Symbols IL1RN
Host Species Mouse
Immunogen Recombinant Human IL-1RA protein (Arg26-Glu177).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16181, 60582
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.