Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL1RA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595457
Description
Rat IL1RA shares 77.6% and 90.7% amino acid (aa) sequence identity with human and mouse IL1RA, respectively. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL.
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
Specifications
IL1RA | |
Polyclonal | |
Unconjugated | |
IL1RN | |
Anakinra; CD121A; cytokine; D2S1473; DIRA; F630041P17Rik; ICIL-1RA; IL 1 receptor antagonist; IL1 inhibitor; il1 receptor; il-1 receptor; IL1 receptor antagonist; IL1F3; IL1R; il-1r; IL1RA; IL-1RA; IL1-RA; IL-1ra3; IL-1R-alpha; IL1RN; IL-1RN; IL1RN (IL1F3); interleukin 1 receptor antagonist; interleukin-1 receptor; interleukin-1 receptor antagonist; interleukin-1 receptor antagonist protein; interleukin-1 receptor antagonist secretory form; intracellular IL-1 receptor antagonist type II; intracellular interleukin-1 receptor antagonist (icIL-1ra); IRAP; IRAP1; MGC10430; MVCD4; P80; type II interleukin-1 receptor antagonist | |
Rabbit | |
Affinity Chromatography | |
RUO | |
60582 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P25086 | |
IL1RN | |
E.coli-derived rat IL1RA recombinant protein (Position: H27-Q178). | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction