Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL1RAPL1 Antibody (2H3C12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | IL1RAPL1 |
---|---|
Clone | 2H3C12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry |
Description
IL1RAPL1 Monoclonal specifically detects IL1RAPL1 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.Specifications
IL1RAPL1 | |
1 mg/mL | |
Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
IL1R8IL1RAPL, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, Oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1 | |
IL1RAPL1 | |
IgG1 | |
Protein A or G purified |
2H3C12 | |
Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.05% Sodium Azide | |
11141 | |
Purified recombinant fragment of human IL1RAPL1 (AA: 541-694) expressed in E. Coli. | |
Primary | |
80 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title