Learn More
IL1RAPL1 Antibody (MM0353-3R16), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP211648C
Catalog No. NBP211648C
Only null left
Add to Cart
Description
Description
IL1RAPL1 Monoclonal specifically detects IL1RAPL1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
IL1RAPL1
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
IL1RAPL1
Human recombinant IL1RAPL1 (extracellular domain)
0.1 mL
11141
Store at 4C in the dark.
IgG2
Western Blot
MM0353-3R16
Western Blot
IL1R8IL1RAPL, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, Oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1
Mouse
Protein G purified
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.