Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL1RAPL1 Mouse anti-Human, Clone: B-L43, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318110200UL
This item is not returnable.
View return policy
Description
IL1RAPL1 Monoclonal antibody specifically detects IL1RAPL1 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
IL1RAPL1 | |
Monoclonal | |
1 mg/ml | |
Flow Cytometry, ELISA | |
IL1R8IL1RAPL, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, Oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1 | |
Recombinant human IL1RAPL1. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells | |
200 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA | |
B-L43 | |
Unconjugated | |
PBS. Sterile-filtered through 0.22 um. | |
Mouse | |
Affinity purified | |
RUO | |
11141 | |
Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction