Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL1RAPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596244
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596244 Supplier Invitrogen™ Supplier No. PA596244
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SISLTVGEND TGLCYNSKMK YFEKAELSKS KEISCRDIED FLLPTREPEI LWYKECRTKT WRPSIVFKRD TLLIREVRED D; Positive Samples: A-431, Mouse brain, Mouse heart, Rat brain, Rat heart; Cellular Location: Cell membrane, Cell projection, Cytoplasm, Single-pass type I membrane protein, axon, dendrite.

The protein encoded by this gene is a member of the interleukin 1 receptor family and is similar to the interleukin 1 accessory proteins. This protein has an N-terminal signal peptide, three extracellular immunoglobulin Ig-like domains, a transmembrane domain, an intracellular Toll/IL-1R domain, and a long C-terminal tail which interacts with multiple signaling molecules. This gene is located at a region on chromosome X that is associated with a non-syndromic form of X-linked intellectual disability. Deletions and mutations in this gene were found in patients with intellectual disability. This gene is expressed at a high level in post-natal brain structures involved in the hippocampal memory system, which suggests a specialized role in the physiological processes underlying memory and learning abilities, and plays a role in synapse formation and stabilization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1RAPL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Il1rapl1
Gene Accession No. P59823, P59824, Q9NZN1
Gene Alias 6330532G10Rik; C030039N24; IL1R8; Il1rapl; Il1rapl1; IL-1-RAPL-1; IL-1RAPL-1; IL1RAPL-1; interleukin 1 receptor accessory protein like 1; interleukin 1 receptor accessory protein-like 1; interleukin 1 receptor-8; interleukin-1 receptor accessory protein-like 1; mental retardation, X-linked 10; MRX10; MRX21; MRX34; Oligophrenin-4; OPHN4; Three immunoglobulin domain-containing IL-1 receptor-related 2; TIGIRR-2; X-linked interleukin-1 receptor accessory protein-like 1
Gene Symbols Il1rapl1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 130-210 of human IL1RAPL1 (NP_0550861).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11141, 317553, 331461
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.