Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL1RAPL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597867
Description
IL1RAPL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The protein encoded by this gene is a member of the interleukin 1 receptor family and is similar to the interleukin 1 accessory proteins. This protein has an N-terminal signal peptide, three extracellular immunoglobulin Ig-like domains, a transmembrane domain, an intracellular Toll/IL-1R domain, and a long C-terminal tail which interacts with multiple signaling molecules. This gene is located at a region on chromosome X that is associated with a non-syndromic form of X-linked intellectual disability. Deletions and mutations in this gene were found in patients with intellectual disability. This gene is expressed at a high level in post-natal brain structures involved in the hippocampal memory system, which suggests a specialized role in the physiological processes underlying memory and learning abilities, and plays a role in synapse formation and stabilization.
Specifications
IL1RAPL1 | |
Polyclonal | |
Unconjugated | |
Il1rapl1 | |
6330532G10Rik; C030039N24; IL1R8; Il1rapl; Il1rapl1; IL-1-RAPL-1; IL-1RAPL-1; IL1RAPL-1; interleukin 1 receptor accessory protein like 1; interleukin 1 receptor accessory protein-like 1; interleukin 1 receptor-8; interleukin-1 receptor accessory protein-like 1; mental retardation, X-linked 10; MRX10; MRX21; MRX34; Oligophrenin-4; OPHN4; Three immunoglobulin domain-containing IL-1 receptor-related 2; TIGIRR-2; X-linked interleukin-1 receptor accessory protein-like 1 | |
Rabbit | |
Protein G | |
RUO | |
11141 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
3.39 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9NZN1 | |
Il1rapl1 | |
Recombinant Human Interleukin-1 receptor accessory protein-like 1 protein (564-679AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction