Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL1RAPL2 Antibody (02), DyLight 594 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305893DL594
Description
IL1RAPL2 Monoclonal antibody specifically detects IL1RAPL2 in Human samples. It is validated for ELISASpecifications
IL1RAPL2 | |
Monoclonal | |
DyLight 594 | |
IL-1R-9, IL1R9IL-1 receptor accessory protein-like 2, IL-1R9IL-1RAPL-2, IL-1-RAPL-2, IL1RAPL-2IL1RAPL-2-related protein, interleukin 1 receptor 9, interleukin 1 receptor accessory protein-like 2, Three immunoglobulin domain-containing IL-1 receptor-related 1, TIGIRR-1Interleukin-1 receptor 9, X-linked interleukin-1 receptor accessory protein-like 2 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL1RAPL2 (NP_059112.1; Met1-Glu356). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
02 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
26280 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction